Dimethylamine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
SIAL295280-100GEA
278
GBP
SIAL295280-100G
SIAL295280-400G
SIAL295280-1KG
Dimethylamine, Sigma-Aldrich®
Dimethylamine
Formula:
C₂H₇N MW: 45.08 g/mol Boiling Pt: 6.9 °C (1013 hPa) Melting Pt: –92.8 °C Density: 0.938 g/cm³ (20 °C) Flash Pt: –55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008288 CAS Number: 124-40-3 EINECS: 204-697-4 UN: 1032 ADR: 2, Merck Index: 12,03278 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...