Diethylamine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
471216-100ML
471216-1L
471216-500ML
471216-5ML
SIAL471216-100MLEA
58.2
GBP
SIAL471216-100ML
SIAL471216-1L
SIAL471216-500ML
SIAL471216-5ML
Diethylamine, Sigma-Aldrich®
Diethylamine
Formula:
(C₂H₅)₂NH Boiling Pt: 55.5 °C (1013 hPa) Melting Pt: –50 °C Density: 0.7048 g/cm³ (20 °C) Flash Pt: –39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009032 CAS Number: 109-89-7 EINECS: 203-716-3 UN: 1154 ADR: 3,II Merck Index: 13,03138 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...