1,8-Diaminooctane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1,8-Octanediamine, Octamethylendiamine

M-Clarity™ quality level = MQ100

D22401-100G D22401-25G
SIALD22401-100GEA 136 GBP
SIALD22401-100G SIALD22401-25G
1,8-Diaminooctane, Sigma-Aldrich®
1,8-Diaminooctane
Formula: C₈H₂₀N₂
MW: 144.26 g/mol
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 51 °C
Density: 0.83 g/cm³ (60 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008248
CAS Number: 373-44-4
EINECS: 206-764-3
UN: 3259
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR