Taurine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Aminoethanesulphonic acid
M-Clarity™ quality level = MQ200
T8691-100G
T8691-1KG
T8691-25G
SIALT8691-100GEA
218
GBP
SIALT8691-100G
SIALT8691-1KG
SIALT8691-25G
Taurine, Sigma-Aldrich®
Taurine
Formula:
C₂H₇NO₃S MW: 125.15 g/mol Melting Pt: 300 Storage Temperature: Ambient |
MDL Number:
MFCD00008197 CAS Number: 107-35-7 EINECS: 203-483-8 Merck Index: 13,09163 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...