Taurine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Aminoethanesulphonic acid

M-Clarity™ quality level = MQ200

T0625-10G T0625-100G T0625-10MG T0625-1KG T0625-25G T0625-500G T-0625
SIALT0625-10GEA 63.5 GBP
SIALT0625-10G SIALT0625-100G SIALT0625-10MG SIALT0625-1KG SIALT0625-25G SIALT0625-500G PFIZT-0625
Taurine, Sigma-Aldrich®
Taurine
Formula: C₂H₇NO₃S
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00008197
CAS Number: 107-35-7
EINECS: 203-483-8
Merck Index: 13,09163

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR