Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ200

T6687-100G T6687-10KG T6687-25KG T6687-5KG
SIALT6687-100GEA 99 GBP
SIALT6687-100G SIALT6687-10KG SIALT6687-25KG SIALT6687-5KG
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Sigma-Aldrich®
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Formula: NH₂C(CH₂OH)₃
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 169…172 °C
Density: 1.328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR