1-Aminobutan-2-ol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002496-5G
002496-1G
FLUO002496-5GEA
0
GBP
FLUO002496-5G
FLUO002496-1G
1-Aminobutan-2-ol
1-Aminobutan-2-ol
Formula:
C₄H₁₁NO |
MDL Number:
MFCD00059196 CAS Number: 13552-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...