Pentylamine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Amylamine, 1-Aminopentane, n-Amylamine, n-Pentylamine, 1-Pentylamine

M-Clarity™ quality level = MQ100

171409-25ML 171409-250ML 171409-50ML
SIAL171409-25MLEA 61.9 GBP
SIAL171409-25ML SIAL171409-250ML SIAL171409-50ML
Pentylamine, Sigma-Aldrich®
Pentylamine
Formula: H₃C(CH₂)₃CH₂NH₂
MW: 87.16 g/mol
Boiling Pt: 104.4
Melting Pt: –50 °C
Density: 0.7547 g/cm³ (20 °C)
Flash Pt: 7
Storage Temperature: Ambient
MDL Number: MFCD00008236
CAS Number: 110-58-7
EINECS: 203-780-2
UN: 1106
ADR: 3 II
Merck Index: 13,00602

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR