Hexadecylamine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
1-Hexadecylamine, n-Hexadecylamine, 1-Aminohexadecane
M-Clarity™ quality level = MQ100
H7408-100G
H7408-500G
SIALH7408-100GEA
55.3
GBP
SIALH7408-100G
SIALH7408-500G
Hexadecylamine, Sigma-Aldrich®
Hexadecylamine
Formula:
H₃C(CH₂)₁₄CH₂NH₂ Boiling Pt: 330 °C (1013 hPa) Melting Pt: 43…46 °C Density: 0.7821 g/cm³ (60 °C) Flash Pt: 140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008158 CAS Number: 143-27-1 EINECS: 205-596-8 UN: 3259 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...