1-Propyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009571-10G
009571-50G
FLUO009571-10GEA
0
GBP
FLUO009571-10G
FLUO009571-50G
1-Propyl isothiocyanate
1-Propyl isothiocyanate
Formula:
C₄H₇NS Boiling Pt: 152…153 °C (1013 hPa) Density: 0,9909 g/cm³ (0 °C) |
MDL Number:
MFCD00004823 CAS Number: 628-30-8 EINECS: 211-035-8 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...