1-Pentyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009557-5G 009557-25G
FLUO009557-5GEA 0 GBP
FLUO009557-5G FLUO009557-25G
1-Pentyl isothiocyanate
1-Pentyl isothiocyanate
Formula: C₆H₁₁NS
Boiling Pt: 191 °C (1013 hPa)
Density: 0,93 g/cm³ (20 °C)
MDL Number: MFCD00014444
CAS Number: 629-12-9
EINECS: 211-075-6
UN: 2922
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR