1-Dodecyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008926-1G
008926-5G
008926-25G
FLUO008926-1GEA
0
GBP
FLUO008926-1G
FLUO008926-5G
FLUO008926-25G
1-Dodecyl isothiocyanate
1-Dodecyl isothiocyanate
Formula:
C₁₃H₂₅NS Boiling Pt: 98…104 °C (0,01 mmHg) Melting Pt: –12 °C Density: 0,8972 g/cm³ (20 °C) |
MDL Number:
MFCD00041121 CAS Number: 1072-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...