1-Decyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Decyl isothiocyanate
008787-5G
008787-1G
FLUO008787-5GEA
0
GBP
FLUO008787-5G
FLUO008787-1G
1-Decyl isothiocyanate
1-Decyl isothiocyanate
Formula:
C₁₁H₂₁NS MW: 199.36 g/mol Boiling Pt: 170…171 °C (35 mmHg) Density: 0,89 g/cm³ (20 °C) |
MDL Number:
MFCD00041120 CAS Number: 24540-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...