Trioctylamine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Tri-n-octylamine

M-Clarity™ quality level = MQ200

T81000-100G T81000-25G T81000-500G
SIALT81000-100GEA 117 GBP
SIALT81000-100G SIALT81000-25G SIALT81000-500G
Trioctylamine, Sigma-Aldrich®
Trioctylamine
Formula: C₂₄H₅₁N
MW: 353.68 g/mol
Boiling Pt: 365
Melting Pt: –39 °C
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 158
Storage Temperature: Ambient
MDL Number: MFCD00009560
CAS Number: 1116-76-3
EINECS: 214-242-1
UN: 3082
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR