1-Iodopentane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Pentyl iodide
M-Clarity™ quality level = MQ100
241946-100G
241946-25G
SIAL241946-100GEA
92.8
GBP
SIAL241946-100G
SIAL241946-25G
1-Iodopentane, Sigma-Aldrich®
1-Iodopentane
Formula:
C₅H₁₁I MW: 198.05 g/mol Boiling Pt: 157 °C (1013 hPa) Melting Pt: –85.6 °C Density: 1.51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001100 CAS Number: 628-17-1 EINECS: 211-030-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...