1-Iodohexane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Hexyl iodide
M-Clarity™ quality level = MQ200
238287-100G
238287-25G
SIAL238287-100GEA
121
GBP
SIAL238287-100G
SIAL238287-25G
1-Iodohexane, Sigma-Aldrich®
1-Iodohexane
Formula:
C₆H₁₃I Boiling Pt: 181 °C (1013 hPa) Melting Pt: –68 °C Density: 1.44 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001102 CAS Number: 638-45-9 EINECS: 211-339-0 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...