1,4-Diiodobutane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Tetramethylene diiodide
M-Clarity™ quality level = MQ100
D122602-100G
D122602-25G
SIALD122602-100GEA
535
GBP
SIALD122602-100G
SIALD122602-25G
1,4-Diiodobutane, Sigma-Aldrich®
1,4-Diiodobutane
Formula:
C₄H₈I₂ MW: 309.92 g/mol Boiling Pt: 247 °C (1013 hPa) Melting Pt: 5.8 °C Density: 2.35 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001099 CAS Number: 628-21-7 EINECS: 211-032-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...