Trifluoromethanesulphonic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
347817-5G
347817-25G
SIAL347817-5GEA
50.2
GBP
SIAL347817-5G
SIAL347817-25G
Trifluoromethanesulphonic acid, Sigma-Aldrich®
Trifluoromethanesulphonic acid
Formula:
CHF₃O₃S Boiling Pt: 162 °C (1013 hPa) Melting Pt: 35 °C Density: 1.69 g/cm³ (20 °C) |
MDL Number:
MFCD00007514 CAS Number: 1493-13-6 EINECS: 216-087-5 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...