Trifluoromethanesulphonic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003328-100G 003328-500G 003328-1KG
FLUO003328-100GEA 0 GBP
FLUO003328-100G FLUO003328-500G FLUO003328-1KG
Trifluoromethanesulphonic acid
Trifluoromethanesulphonic acid
Formula: CHF₃O₃S
Boiling Pt: 162 °C (1013 hPa)
Melting Pt: 35 °C
Density: 1,69 g/cm³ (20 °C)
MDL Number: MFCD00007514
CAS Number: 1493-13-6
EINECS: 216-087-5
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR