Trifluoromethanesulphonic anhydride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

007526-50G 007526-250G
FLUO007526-50GEA 0 GBP
FLUO007526-50G FLUO007526-250G
Trifluoromethanesulphonic anhydride
Trifluoromethanesulphonic anhydride
Formula: C₂F₆O₅S₂
Boiling Pt: 83 °C (1013 hPa)
Density: 1,67 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000408
CAS Number: 358-23-6
EINECS: 206-616-8
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR