Triisopropylsilyl trifluoromethanesulphonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009411-5G 009411-25G
FLUO009411-5GEA 0 GBP
FLUO009411-5G FLUO009411-25G
Triisopropylsilyl trifluoromethanesulphonate
Triisopropylsilyl trifluoromethanesulphonate
Formula: C₁₀H₂₁F₃O₃SSi
Boiling Pt: 120 °C (1013 hPa)
Density: 1,136 g/cm³ (20 °C)
MDL Number: MFCD00009913
CAS Number: 80522-42-5
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR