4,4,4-Trifluoro-1-butanol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
001532-1G
001532-5G
001532-25G
FLUO001532-1GEA
0
GBP
FLUO001532-1G
FLUO001532-5G
FLUO001532-25G
4,4,4-Trifluoro-1-butanol
4,4,4-Trifluoro-1-butanol
Formula:
C₄H₇F₃O Boiling Pt: 122 °C (1013 hPa) Density: 1,217 g/cm³ (25 °C) |
MDL Number:
MFCD00041483 CAS Number: 461-18-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...