2,2-Difluoroethanol ≥97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

B22201.03 B22201.06
B22201.03EA 32.4 GBP
B22201.03 B22201.06
2,2-Difluoroethanol ≥97%
2,2-Difluoroethanol

Purity: 97% CAS No.: 359-13-7 Molecular Formula: HCF2CH2OH Formula Weight: 82.05 Flash Point: 3°(37°F) Refractive Index: 1.3335 Melting Point: -28° Boiling Point: 95-96° Beilstein Reference: 1731617 UN No.: 1987 MDL No.: MFCD00042248

Formula: C₂H₄F₂O
MW: 82.05 g/mol
Boiling Pt: 95…96 °C
Melting Pt: –28 °C
Density: 1.296
Flash Pt: 3 °C (37 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042248
CAS Number: 359-13-7
UN: 1987
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR