2,2,3,4,4,4-Hexafluoro-1-butanol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002398-5G 002398-100G 002398-25G
FLUO002398-5GEA 0 GBP
FLUO002398-5G FLUO002398-100G FLUO002398-25G
2,2,3,4,4,4-Hexafluoro-1-butanol
2,2,3,4,4,4-Hexafluoro-1-butanol
Formula: C₄H₄F₆O
Boiling Pt: 104…105 °C (1013 hPa)
Density: 1,564 g/cm³ (20 °C)
MDL Number: MFCD00042309
CAS Number: 382-31-0
EINECS: 206-842-7
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR