2,2,3,3,4,4,4-Heptafluoro-1-butanol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
001237-100G
001237-25G
FLUO001237-100GEA
0
GBP
FLUO001237-100G
FLUO001237-25G
2,2,3,3,4,4,4-Heptafluoro-1-butanol
2,2,3,3,4,4,4-Heptafluoro-1-butanol
Formula:
C₄H₃F₇O Boiling Pt: 96 °C (1013 hPa) Density: 1,6 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004674 CAS Number: 375-01-9 EINECS: 206-782-1 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...