1,1,1-Trifluoro-2-propanol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001252-5G 001252-25G
FLUO001252-5GEA 0 GBP
FLUO001252-5G FLUO001252-25G
1,1,1-Trifluoro-2-propanol
1,1,1-Trifluoro-2-propanol
Formula: C₃H₅F₃O
Boiling Pt: 75…76 °C (1013 hPa)
Melting Pt: –52 °C
Density: 1,259 g/cm³ (20 °C)
MDL Number: MFCD00014403
CAS Number: 374-01-6
EINECS: 206-773-2
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR