1,1,1,3,3,3-Hexafluoro-2-propanol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP

024830-250G 024830-50G
FLUO024830-250GEA 0 GBP
FLUO024830-250G FLUO024830-50G
1,1,1,3,3,3-Hexafluoro-2-propanol
1,1,1,3,3,3-Hexafluoro-2-propanol
Formula: C₃H₂F₆O
Boiling Pt: 59.1 °C (1013 hPa)
Melting Pt: –96 °C
Density: 1.492 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011651
CAS Number: 920-66-1
EINECS: 213-059-4
UN: 1760
ADR: 8,II
Merck Index: 13,09598

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR