Perfluoro-tert-butanol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003281-5G 003281-25G 003281-100G
FLUO003281-5GEA 0 GBP
FLUO003281-5G FLUO003281-25G FLUO003281-100G
Perfluoro-tert-butanol
Perfluoro-tert-butanol
Formula: C₄HF₉O
Boiling Pt: 84 °C (1013 hPa)
Melting Pt: –17 °C
Density: 1,693 g/cm³ (20 °C)
MDL Number: MFCD00042092
CAS Number: 2378-02-1
EINECS: 219-157-3
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR