Trifluoroacetic anhydride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Trifluoracetic anhydride, TFAA
001272-100G
001272-500G
001272-1KG
FLUO001272-100GEA
0
GBP
FLUO001272-100G
FLUO001272-500G
FLUO001272-1KG
Trifluoroacetic anhydride
Trifluoroacetic anhydride
Formula:
C₄F₆O₃ Boiling Pt: 39.4 °C (1013 hPa) Melting Pt: –65 °C Density: 1,487 g/cm³ (20 °C) |
MDL Number:
MFCD00000416 CAS Number: 407-25-0 EINECS: 206-982-9 UN: 3265 ADR: 8,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...