Pentafluoropropionic anhydride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Perfluoropropanoic anhydride, Pentafluoropropanoic anhydride, PFPA
003277-25G
003277-100G
FLUO003277-25GEA
0
GBP
FLUO003277-25G
FLUO003277-100G
Pentafluoropropionic anhydride
Pentafluoropropionic anhydride
Formula:
C₆F₁₀O₃ MW: 310.05 g/mol Boiling Pt: 69 Density: 1.571 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000429 CAS Number: 356-42-3 EINECS: 206-604-2 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...