2-Iodoperfluoropropane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,1,1,2,3,3,3-Heptafluoro-2-iodopropane, Heptafluoroisopropyl iodide, Heptafluoro-2-iodopropane, 2-Iodoheptafluoropropane

003260-25G 003260-100G 003260-10G
FLUO003260-25GEA 0 GBP
FLUO003260-25G FLUO003260-100G FLUO003260-10G
2-Iodoperfluoropropane
2-Iodoperfluoropropane
Formula: C₃F₇I
MW: 295.93 g/mol
Boiling Pt: 39
Melting Pt: –56 °C
Density: 2.077 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042089
CAS Number: 677-69-0
EINECS: 211-643-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR