1-Iodoperfluorbutane
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
|
Synonyms:
Perfluorobutyl iodide, 1,1,1,2,2,3,3,4,4-Nonafluoro-4-iodobutane, 1-Iodononafluorobutane, Nonafluoro-1-iodobutane
002874-25G
002874-100G
002874-5G
FLUO002874-25GEA
0
GBP
FLUO002874-25G
FLUO002874-100G
FLUO002874-5G
1-Iodoperfluorbutane
1-Iodoperfluorbutane
Formula:
C₄F₉I MW: 345.93 g/mol Boiling Pt: 65 °C (1013 hPa) Melting Pt: –2 °C Density: 2,0497 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001062 CAS Number: 423-39-2 EINECS: 207-025-8 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...