Tetrafluorosuccinic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2,2,3,3-Tetrafluorosuccinic acid, Tetrafluoro-1,4-butanedioic acid
002270-5G
002270-25G
FLUO002270-5GEA
0
GBP
FLUO002270-5G
FLUO002270-25G
Tetrafluorosuccinic acid
Tetrafluorosuccinic acid
Formula:
C₄H₂F₄O₄ MW: 190.05 g/mol Boiling Pt: 120 °C (1 mmHg) Melting Pt: 118…120 °C Density: 1,4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042430 CAS Number: 377-38-8 EINECS: 206-820-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...