Hexafluoroglutaric anhydride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
003252-5G
003252-25G
003252-100G
FLUO003252-5GEA
0
GBP
FLUO003252-5G
FLUO003252-25G
FLUO003252-100G
Hexafluoroglutaric anhydride
Hexafluoroglutaric anhydride
Formula:
C₅F₆O₃ Boiling Pt: 80 °C (1013 hPa) Density: 1,7231 g/cm³ (0 °C) |
MDL Number:
MFCD00006680 CAS Number: 376-68-1 EINECS: 206-811-8 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...