Hexafluoroglutaric acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Hexafluoro-1,5-pentanedioic acid

002271-5G 002271-25G 002271-100G
FLUO002271-5GEA 0 GBP
FLUO002271-5G FLUO002271-25G FLUO002271-100G
Hexafluoroglutaric acid
Hexafluoroglutaric acid
Formula: C₅H₂F₆O₄
Boiling Pt: 134…138 °C (3 mmHg)
Melting Pt: 92…95 °C
MDL Number: MFCD00004172
CAS Number: 376-73-8
EINECS: 206-813-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR