Pentafluoropropionic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Perfluoropropionic acid, Pentafluoropropanoic acid, Perfluoropropanoic acid, 2,2,3,3,3-Pentafluoropropionic acid
002471-25G
002471-100G
002471-500G
FLUO002471-25GEA
0
GBP
FLUO002471-25G
FLUO002471-100G
FLUO002471-500G
Pentafluoropropionic acid
Pentafluoropropionic acid
Formula:
C₃HF₅O₂ MW: 164.03 g/mol Boiling Pt: 96 Density: 1.561 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004170 CAS Number: 422-64-0 EINECS: 207-021-6 UN: 3265 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...