Heptafluorobutyric acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Perfluorobutyric acid, HFBA, Heptafluorobutanoic acid

001269-25G 001269-100G
FLUO001269-25GEA 0 GBP
FLUO001269-25G FLUO001269-100G
Heptafluorobutyric acid
Heptafluorobutyric acid
Formula: C₄HF₇O₂
Boiling Pt: 121 °C (1013 hPa)
Melting Pt: –17.5 °C
Density: 1,764 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004171
CAS Number: 375-22-4
EINECS: 206-786-3
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR