Difluoroacetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

001231-25G 001231-100G
FLUO001231-25GEA 0 GBP
FLUO001231-25G FLUO001231-100G
Difluoroacetic acid
Difluoroacetic acid
Formula: C₂H₂F₂O₂
Boiling Pt: 134 °C (1013 hPa)
Melting Pt: –1 °C
Density: 1,526 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004220
CAS Number: 381-73-7
EINECS: 206-839-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR