Difluoroacetic acid ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
A12787.06
A12787.14
A12787.06EA
52.4
GBP
A12787.06
A12787.14
Difluoroacetic acid ≥98%
Difluoroacetic acid
Purity: 98%
CAS No.: 381-73-7
Molecular Formula: F2CHCO2H
Formula Weight: 96.03
Flash Point: 78°(172°F)
Refractive Index: 1.3430
Odour: Pungent
Melting Point: -1°
Boiling Point: 132-134°
Beilstein Reference: 1098588
UN No.: 3265
MDL No.: MFCD00004220
Formula:
C₂H₂F₂O₂ MW: 96.03 g/mol Boiling Pt: 132…134 °C Melting Pt: –1 °C Density: 1.526 Flash Pt: 78 °C (172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004220 CAS Number: 381-73-7 EINECS: 206-839-0 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...