Ethyl trifluoroacetate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Trifluoroacetic ethyl ester

001179-100G 001179-500G 001179-1KG
FLUO001179-100GEA 0 GBP
FLUO001179-100G FLUO001179-500G FLUO001179-1KG
Ethyl trifluoroacetate
Ethyl trifluoroacetate
Formula: C₄H₅F₃O₂
Boiling Pt: 61 °C (1013 hPa)
Melting Pt: –78 °C
Density: 1,19 g/cm³ (20 °C)
Flash Pt: –17 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000419
CAS Number: 383-63-1
EINECS: 206-851-6
UN: 2924
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR