Ethyl fluoroacetate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Fluoroacetic ethyl ester

019465-100G 019465-25G
FLUO019465-100GEA 0 GBP
FLUO019465-100G FLUO019465-25G
Ethyl fluoroacetate
Ethyl fluoroacetate
Formula: C₄H₇FO₂
Boiling Pt: 119.2 °C (1013 hPa)
Density: 1,0912 g/cm³ (20 °C)
MDL Number: MFCD00000450
CAS Number: 459-72-3
EINECS: 207-297-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR