Methyl difluoroacetate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
006791-1G
006791-5G
006791-25G
FLUO006791-1GEA
0
GBP
FLUO006791-1G
FLUO006791-5G
FLUO006791-25G
Methyl difluoroacetate
Methyl difluoroacetate
Formula:
C₃H₄F₂O₂ Boiling Pt: 84…85 °C (1013 hPa) Density: 1,272 g/cm³ (20 °C) |
MDL Number:
MFCD00039276 CAS Number: 433-53-4 EINECS: 207-089-7 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...