1,1,1-Trifluoroacetylacetone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1,1,1-Trifluoropentane-2,4-dione, 1,1,1-Trifluoro-2,4-pentanedione

M-Clarity™ quality level = MQ100

235970-10G 235970-25G
SIAL235970-10GEA 118 GBP
SIAL235970-10G SIAL235970-25G
1,1,1-Trifluoroacetylacetone, Sigma-Aldrich®
1,1,1-Trifluoroacetylacetone
Formula: C₅H₅F₃O₂
MW: 154.09 g/mol
Boiling Pt: 107
Density: 1.28 g/cm³ (20 °C)
Flash Pt: 28
Storage Temperature: Refrigerator
MDL Number: MFCD00000427
CAS Number: 367-57-7
EINECS: 206-698-5
UN: 1224
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR