Thiourea, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Thiocarbamide, Sulphourea

M-Clarity™ quality level = MQ200

T8656-100G T8656-500G T8656-50G
SIALT8656-100GEA 67.6 GBP
SIALT8656-100G SIALT8656-500G SIALT8656-50G
Thiourea, Sigma-Aldrich®
Thiourea
Formula: NH₂CSNH₂
MW: 76.12 g/mol
Melting Pt: 174…177 °C
Density: 1.435 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008067
CAS Number: 62-56-6
EINECS: 200-543-5
UN: 3077
ADR: 9,III
Merck Index: 13,09443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&vid=e9ae5dc49e92d68f893244a5cd4e0245f289ed09&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR