Thiourea, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Thiocarbamide, Sulphourea
T7875-5KG
T7875-100G
T7875-500G
T7875-5G
T7875-1KG
SIALT7875-5KGEA
426
GBP
SIALT7875-5KG
SIALT7875-100G
SIALT7875-500G
SIALT7875-5G
SIALT7875-1KG
Thiourea, Sigma-Aldrich®
Thiourea
Formula:
NH₂CSNH₂ MW: 76.12 g/mol Melting Pt: 174…177 °C Density: 1.435 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008067 CAS Number: 62-56-6 EINECS: 200-543-5 UN: 3077 ADR: 9,III Merck Index: 13,09443 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...