Thioacetic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ300

W421001-100G W421001-1KG W421001-SAMPLE
SIALW421001-100GEA 127 GBP
SIALW421001-100G SIALW421001-1KG SIALW421001-SAMPLE
Thioacetic acid, Sigma-Aldrich®
Thioacetic acid
Formula: C₂H₄OS
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –17 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 18 °C
MDL Number: MFCD00004853
CAS Number: 507-09-5
EINECS: 208-063-8
UN: 2436
ADR: 3,II
Merck Index: 12,09454

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR