Suberic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1,8-Octanedioic acid, Octanedioic acid, 1,6-Hexanedicarboxylic acid
M-Clarity™ quality level = MQ100
S5200-100G
S5200-500G
S5200-5G
SIALS5200-100GEA
63.5
GBP
SIALS5200-100G
SIALS5200-500G
SIALS5200-5G
Suberic acid, Sigma-Aldrich®
Suberic acid
Formula:
C₈H₁₄O₄ Boiling Pt: 361 °C (1013 hPa) Melting Pt: 139…142 °C Density: 1.354 g/cm³ (21 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004428 CAS Number: 505-48-6 EINECS: 208-010-9 Merck Index: 12,09031 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...