Glycine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Aminoacetic acid, Glycocoll, H-Gly-OH
M-Clarity™ quality level = MQ200
33226-1KG
33226-250G
33226-50KG
SIAL33226-1KGEA
169
GBP
SIAL33226-1KG
SIAL33226-250G
SIAL33226-50KG
Glycine, Sigma-Aldrich®
Glycine
Formula:
NH₂CH₂COOH MW: 75.07 g/mol Boiling Pt: 181 Melting Pt: 83 Density: 1.64 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008131 CAS Number: 56-40-6 EINECS: 200-272-2 Merck Index: 13,04504 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...