Zinc stearate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Stearic acid zinc salt, Zinc(II) stearate

M-Clarity™ quality level = MQ100

26423-1KG 26423-10KG
SIAL26423-1KGEA 83.8 GBP
SIAL26423-1KG SIAL26423-10KG
Zinc stearate, Sigma-Aldrich®
Zinc stearate
Formula: C₃₆H₇₀O₄Zn
MW: 632.34 g/mol
Melting Pt: 140
Density: 1.095 g/cm³ (20 °C)
Flash Pt: 276
Storage Temperature: Ambient
MDL Number: MFCD00013031
CAS Number: 557-05-1
EINECS: 209-151-9
Merck Index: 13,10212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8ad7f3e101681798edbd54e57ad5d2ac82b2e5d&vid=435308cdcb35862fe6cc06384da2ef10d84c5700&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR