Sodium propionate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Propionic acid sodium salt, Sodiumpropionate
M-Clarity™ quality level = MQ200
P1880-100G
P1880-1KG
P1880-500G
SIALP1880-100GEA
41.2
GBP
SIALP1880-100G
SIALP1880-1KG
SIALP1880-500G
Sodium propionate, Sigma-Aldrich®
Sodium propionate
Formula:
C₃H₅NaO₂ MW: 96.06 g/mol Melting Pt: 288.76 Flash Pt: 157 Storage Temperature: Ambient |
MDL Number:
MFCD00002759 CAS Number: 137-40-6 EINECS: 205-290-4 Merck Index: 12,08816 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...